missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethylbenzimidazole 99.0+%, TCI America™

Catalog No. D134525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D134525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D134525G Supplier TCI America Supplier No. D134525G

Chemical Identifiers

CAS 582-60-5
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005603
InChI Key LJUQGASMPRMWIW-UHFFFAOYSA-N
Synonym 5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole
PubChem CID 675
ChEBI CHEBI:15890
IUPAC Name 5,6-dimethyl-1H-1,3-benzodiazole
SMILES CC1=C(C)C=C2N=CNC2=C1

Specifications

Melting Point 205°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethylbenzimidazole
RTECSNumber DD8280000
TSCA Yes