missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethylbenzimidazole, 99+%

Catalog No. p-116432
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115980250 25 g Glass bottle
AC115981000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115980250 Supplier Thermo Scientific Chemicals Supplier No. 115980250

CAS: 582-60-5 | C9H10N2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 582-60-5
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00005603
InChI Key LJUQGASMPRMWIW-UHFFFAOYSA-N
Synonym 5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole
PubChem CID 675
ChEBI CHEBI:15890
IUPAC Name 5,6-dimethyl-1H-1,3-benzodiazole
SMILES CC1=C(C)C=C2N=CNC2=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 203°C to 206°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 3258
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in diluted acids,soluble in chloroform,ether and ethanol
Formula Weight 146.19
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5, 6-Dimethylbenzimidazole, 99+%
Show More Show Less
EINECSNumber 209-488-1
RTECSNumber DD8280000
TSCA TSCA

RUO – Research Use Only