missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-2-benzimidazolinone, 98%

Catalog No. p-7022577
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1542103 1 g
AAA1542106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1542103 Supplier Thermo Scientific Chemicals Supplier No. A1542103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2033-30-9 | C9H10N2O | 162.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2033-30-9
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.192
MDL Number MFCD00051725
InChI Key ORWJLFLEIZBRBR-UHFFFAOYSA-N
Synonym 5,6-dimethyl-1h-benzo d imidazol-2 3h-one, 5,6-dimethyl-2-benzimidazolinone, 1,3-dihydro-5,6-dimethyl-2h-benzimidazol-2-one, 5,6-dimethylbenzimidazolin-2-one, 2h-benzimidazol-2-one, 1,3-dihydro-5,6-dimethyl, 2h-benzimidazol-2-one,1,3-dihydro-5,6-dimethyl, 5,6-dimethyl-3-hydrobenzimidazol-2-one, 5,6-dimethyl-1,3-dihydro-1,3-benzodiazol-2-one, 5,6-dimethyl-1,3-dihydro-benzoimidazol-2-one, acmc-1cmtt
PubChem CID 74853
IUPAC Name 5,6-dimethyl-1,3-dihydrobenzimidazol-2-one
SMILES CC1=CC2=C(C=C1C)NC(=O)N2

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 139589
Formula Weight 162.19
Percent Purity 98%
Chemical Name or Material 5,6-Dimethyl-2-benzimidazolinone
EINECSNumber 217-993-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only