missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dimethoxyindole 98.0+%, TCI America™
Supplier: TCI America D51481G
Specifications
| 5,6-Dimethoxyindole | |
| 155°C | |
| C10H11NO2 | |
| 1 g | |
| QODBZRNBPUPLEZ-UHFFFAOYSA-N | |
| 5,6-dimethoxy-1H-indole | |
| 84431 | |
| ≥98.0% (GC) |
| 14430-23-0 | |
| 198°C | |
| MFCD00005675 | |
| 5,6-dimethoxyindole, 1h-indole, 5,6-dimethoxy, 5,6-dimethoxyindol, zlchem 178, pubchem7329, acmc-1cbhq, 1h-indole,5,6-dimethoxy, # | |
| COC1=C(OC)C=C2C=CNC2=C1 | |
| 177.20 | |
| 177.20 | |
| Crystalline Powder |
Chemical Identifiers
| 14430-23-0 | |
| 177.20 | |
| QODBZRNBPUPLEZ-UHFFFAOYSA-N | |
| 84431 | |
| COC1=C(OC)C=C2C=CNC2=C1 |
| C10H11NO2 | |
| MFCD00005675 | |
| 5,6-dimethoxyindole, 1h-indole, 5,6-dimethoxy, 5,6-dimethoxyindol, zlchem 178, pubchem7329, acmc-1cbhq, 1h-indole,5,6-dimethoxy, # | |
| 5,6-dimethoxy-1H-indole |
Safety and Handling
TSCA : No