missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxyindole, 98%, Thermo Scientific Chemicals

Catalog No. AAB2112603
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2112603 1 g
AAB2112601 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2112603 Supplier Thermo Scientific Chemicals Supplier No. B2112603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

It is applied as a reactant in synthesis of indolylhydroxyoxindoles via enantioselective Friedel-Crafts reaction, in synthesis of benzyl trimethoxyindoles, for synthesis of benzoylpiperazinyl-indolyl ethane dione derivatives as HIV-1 inhibitors, for synthesis of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors and for preparation of tryptophanol derivatives via the Grignard reaction. It is an indole derivative as analgesic anti-inflammatory useful in treatment of pain, inflammation. It is also used in the synthesis of N-benzyl-N-cyclopropyl-5,6-dimethoxyindole-3-glyoxalamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reactant in synthesis of indolylhydroxyoxindoles via enantioselective Friedel-Crafts reaction, in synthesis of benzyl trimethoxyindoles, for synthesis of benzoylpiperazinyl-indolyl ethane dione derivatives as HIV-1 inhibitors, for synthesis of 1-aroylindole 3-aroylindoles combretastatin A-4 analogs as antitumor agents and tubulin polymerization inhibitors and for preparation of tryptophanol derivatives via the Grignard reaction. It is an indole derivative as analgesic anti-inflammatory useful in treatment of pain, inflammation. It is also used in the synthesis of N-benzyl-N-cyclopropyl-5,6-dimethoxyindole-3-glyoxalamide.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 14430-23-0
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00005675
InChI Key QODBZRNBPUPLEZ-UHFFFAOYSA-N
PubChem CID 84431
IUPAC Name 5,6-dimethoxy-1H-indole
SMILES COC1=C(OC)C=C2C=CNC2=C1

Specifications

Melting Point 154°C to 160°C
Boiling Point 198°C (8 mmHg)
Quantity 1 g
Beilstein 5683
Synonym 5,6-dimethoxyindole, 1h-indole, 5,6-dimethoxy, 5,6-dimethoxyindol, zlchem 178, pubchem7329, acmc-1cbhq, 1h-indole,5,6-dimethoxy, #
Solubility Information Insoluble in water.
Formula Weight 177.2
Percent Purity 98%
Chemical Name or Material 5,6-Dimethoxyindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-402-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only