missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxyindole-2-carboxylic acid, 98%

Catalog No. AAL0073306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0073306 5 g
AAL0073303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0073306 Supplier Thermo Scientific Chemicals Supplier No. L0073306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88210-96-2 | C11H11NO4 | 221.21 g/mol

5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 88210-96-2
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.21
MDL Number MFCD00015459
InChI Key UZHQHNDRVITJPL-UHFFFAOYSA-N
Synonym 5,6-dimethoxyindole-2-carboxylic acid, 5,6-dimethoyxindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5,6-dimethoxy, 5,6-dica, 5,6-dimethoxyindole-2-carboxylicacid, 5,6-dimethoxyindolyl-2-carboxylic acid, 1h-indole-2-carboxylicacid, 5,6-dimethoxy
PubChem CID 139035
IUPAC Name 5,6-dimethoxy-1H-indole-2-carboxylic acid
SMILES COC1=C(OC)C=C2C=C(NC2=C1)C(O)=O

Specifications

Melting Point ∽210°C (decomposition)
Quantity 5 g
Beilstein 202249
Solubility Information Insoluble in water.
Formula Weight 221.22
Percent Purity 98%
Chemical Name or Material 5,6-Dimethoxyindole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only