missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dimethoxy-2-(4-piperidylmethyl)-1-indanone Hydrochloride 98.0+%, TCI America™
$76.43 - $382.13
Chemical Identifiers
| CAS | 120013-39-0 |
|---|---|
| Molecular Formula | C17H24ClNO3 |
| Molecular Weight (g/mol) | 325.83 |
| MDL Number | MFCD09033179 |
| InChI Key | WOZXDQQCMZIQEG-UHFFFAOYNA-N |
| PubChem CID | 14553606 |
| IUPAC Name | 5,6-dimethoxy-2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-inden-1-one hydrochloride |
| SMILES | Cl.COC1=C(OC)C=C2C(=O)C(CC3CCNCC3)CC2=C1 |
Chemical Identifiers
| 120013-39-0 | |
| 325.83 | |
| WOZXDQQCMZIQEG-UHFFFAOYNA-N | |
| 5,6-dimethoxy-2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-inden-1-one hydrochloride |
| C17H24ClNO3 | |
| MFCD09033179 | |
| 14553606 | |
| Cl.COC1=C(OC)C=C2C(=O)C(CC3CCNCC3)CC2=C1 |
Specifications
| 120013-39-0 | |
| Yellow | |
| MFCD09033179 | |
| WOZXDQQCMZIQEG-UHFFFAOYNA-N | |
| 5,6-dimethoxy-2-[(piperidin-4-yl)methyl]-2,3-dihydro-1H-inden-1-one hydrochloride | |
| 14553606 | |
| ≥98.0% (HPLC,N) | |
| 5,6-Dimethoxy-2-(4-piperidylmethyl)-1-indanone Hydrochloride |
| 250°C | |
| C17H24ClNO3 | |
| 1 g | |
| Cl.COC1=C(OC)C=C2C(=O)C(CC3CCNCC3)CC2=C1 | |
| 325.83 | |
| 325.83 | |
| Crystalline Powder |
Safety and Handling
TSCA : No