missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxy-1-indanone 98.0+%, TCI America™

Catalog No. D33135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33135G 5 g
D331325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33135G Supplier TCI America Supplier No. D33135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2107-69-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
InChI Key IHMQOBPGHZFGLC-UHFFFAOYSA-N
Synonym 5,6-dimethoxy-1-indanone, 5,6-dimethoxy-2,3-dihydro-1h-inden-1-one, 5,6-dimethoxyindan-1-one, 5,6-dimethoxy-indan-1-one, 1h-inden-1-one, 2,3-dihydro-5,6-dimethoxy, 5,6-dimethoxyindanone, unii-f805rzi8go, f805rzi8go, 1-indanone, 5,6-dimethoxy, 5,6-dimethoxyindan-1-on
PubChem CID 75018
IUPAC Name 5,6-dimethoxy-2,3-dihydroinden-1-one
SMILES COC1=C(C=C2C(=C1)CCC2=O)OC

Specifications

Melting Point 120°C
Boiling Point 139°C
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dimethoxy-1-indanone
EINECSNumber (4)-1806
TSCA Yes