missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxy-1-indanone, 97%

Catalog No. AAB2555906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2555906 5 g
AAB2555914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2555906 Supplier Thermo Scientific Chemicals Supplier No. B2555906

CAS: 2107-69-9 | C11H12O3 | 192.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2107-69-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00003790
InChI Key IHMQOBPGHZFGLC-UHFFFAOYSA-N
Synonym 5,6-dimethoxy-1-indanone, 5,6-dimethoxy-2,3-dihydro-1h-inden-1-one, 5,6-dimethoxyindan-1-one, 5,6-dimethoxy-indan-1-one, 1h-inden-1-one, 2,3-dihydro-5,6-dimethoxy, 5,6-dimethoxyindanone, unii-f805rzi8go, f805rzi8go, 1-indanone, 5,6-dimethoxy, 5,6-dimethoxyindan-1-on
PubChem CID 75018
IUPAC Name 5,6-dimethoxy-2,3-dihydroinden-1-one
SMILES COC1=C(C=C2C(=C1)CCC2=O)OC

Specifications

Melting Point 118°C to 120°C
Quantity 5 g
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material 5,6-Dimethoxy-1-indanone
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 218-287-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only