missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dihydroxyindole, 95%
CAS: 3131-52-0 | C8H7NO2 | 149.149 g/mol
$428.97 - $1601.14
Chemical Identifiers
| CAS | 3131-52-0 |
|---|---|
| Molecular Formula | C8H7NO2 |
| Molecular Weight (g/mol) | 149.149 |
| MDL Number | MFCD00798933 |
| InChI Key | SGNZYJXNUURYCH-UHFFFAOYSA-N |
| Synonym | 5,6-dihydroxyindole, dopamine lutine, 3h-indole-5,6-diol, 5,6-dihydroxy-1h-indole, dhi, unii-z3oc8499kg, 5,6-dihydroxyindolei, chembl92636, 3id, 5,6-dihydroxy indole |
| PubChem CID | 114683 |
| ChEBI | CHEBI:27404 |
| IUPAC Name | 1H-indole-5,6-diol |
| SMILES | C1=CNC2=CC(=C(C=C21)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6670903
|
Thermo Scientific Chemicals
H6670903 |
1 g |
Each for $428.97
|
|
|||||
|
AAH6670906
|
Thermo Scientific Chemicals
H6670906 |
5 g |
Each for $1,601.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3131-52-0 | |
| 149.149 | |
| SGNZYJXNUURYCH-UHFFFAOYSA-N | |
| 114683 | |
| 1H-indole-5,6-diol |
| C8H7NO2 | |
| MFCD00798933 | |
| 5,6-dihydroxyindole, dopamine lutine, 3h-indole-5,6-diol, 5,6-dihydroxy-1h-indole, dhi, unii-z3oc8499kg, 5,6-dihydroxyindolei, chembl92636, 3id, 5,6-dihydroxy indole | |
| CHEBI:27404 | |
| C1=CNC2=CC(=C(C=C21)O)O |
Specifications
| 3131-52-0 | |
| MFCD00798933 | |
| 122055 | |
| SGNZYJXNUURYCH-UHFFFAOYSA-N | |
| 1H-indole-5,6-diol | |
| 114683 | |
| 149.15 | |
| Solid |
| C8H7NO2 | |
| 1 g | |
| 5,6-dihydroxyindole, dopamine lutine, 3h-indole-5,6-diol, 5,6-dihydroxy-1h-indole, dhi, unii-z3oc8499kg, 5,6-dihydroxyindolei, chembl92636, 3id, 5,6-dihydroxy indole | |
| C1=CNC2=CC(=C(C=C21)O)O | |
| 149.149 | |
| CHEBI:27404 | |
| 95% | |
| 5,6-Dihydroxyindole |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 412-130-9
RTECSNumber : NL7858000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only