missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydroxyindole, 95%

Catalog No. AAH6670906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6670906 5 g
AAH6670903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6670906 Supplier Thermo Scientific Chemicals Supplier No. H6670906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3131-52-0 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3131-52-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00798933
InChI Key SGNZYJXNUURYCH-UHFFFAOYSA-N
Synonym 5,6-dihydroxyindole, dopamine lutine, 3h-indole-5,6-diol, 5,6-dihydroxy-1h-indole, dhi, unii-z3oc8499kg, 5,6-dihydroxyindolei, chembl92636, 3id, 5,6-dihydroxy indole
PubChem CID 114683
ChEBI CHEBI:27404
IUPAC Name 1H-indole-5,6-diol
SMILES C1=CNC2=CC(=C(C=C21)O)O

Specifications

Quantity 5 g
Beilstein 122055
Formula Weight 149.15
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5,6-Dihydroxyindole
Hazard Category H302
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 412-130-9
RTECSNumber NL7858000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only