missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydrodicyclopentadiene 95.0+%, TCI America™

Catalog No. D200125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D200125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D200125G Supplier TCI America Supplier No. D200125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4488-57-7
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00078238
InChI Key HANKSFAYJLDDKP-UHFFFAOYSA-N
Synonym 3a,4,5,6,7,7a-Hexahydro-4,7-methano-1H-indene, Tricyclo[5.2.1.0(2,6)]decan-3-ene
PubChem CID 95605
SMILES C1CC2CC1C3C2C=CC3

Specifications

Melting Point 49°C
Color White
Boiling Point 180°C
Quantity 25 g
Formula Weight 134.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dihydrodicyclopentadiene