missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-5-methyluracil, 98+%

Catalog No. p-7043672
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0199606 5 g
AAL0199614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0199606 Supplier Thermo Scientific Chemicals Supplier No. L0199606
Only null left

CAS: 696-04-8 | C5H8N2O2 | 128.13 g/mol

5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,6-Dihydro-5-methyluracil, is used as an active pharmaceutical ingredient.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 696-04-8
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00023159
InChI Key NBAKTGXDIBVZOO-UHFFFAOYNA-N
Synonym dihydrothymine, 5,6-dihydro-5-methyluracil, 5,6-dihydrothymine, 5-methyl-5,6-dihydrouracil, hydrouracil, 5-methyl, 5-methyl-hydrouracil, 5-methyldihydropyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, dihydro-5-methyl, 5,6-dihydro-5-methyl-2,4-dioxypyrimidine, 5,6-dihydro-2,4-dihydroxy-5-methylpyrimidine
PubChem CID 93556
ChEBI CHEBI:27468
IUPAC Name 5-methyl-1,3-diazinane-2,4-dione
SMILES CC1CNC(=O)NC1=O

Specifications

Melting Point 263°C to 265°C
Quantity 5 g
Beilstein 81983
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 128.13
Percent Purity ≥98%
Chemical Name or Material 5,6-Dihydro-5-methyluracil
EINECSNumber 211-787-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only