missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro-2H-pyran-2-one, 90%

Catalog No. AC426640010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426640010 1 g Glass bottle
AC426640050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426640010 Supplier Thermo Scientific Chemicals Supplier No. 426640010

CAS: 3393-45-1 | C5H6O2 | 98.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3393-45-1
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00010439
InChI Key VREFDQNWPNDZEX-UHFFFAOYSA-N
Synonym 5,6-dihydro-2h-pyran-2-one, 2h-pyran-2-one, 5,6-dihydro, 5,6-dihydropyran-2-one, 5,6-dihydro-pyran-2-one, chembl55078, 5,6-dihydro-2-pyranone, acmc-1ck70, 2-pentenoic acid, 5-hydroxy-, .delta.-lactone, 5,6-dihydro-2h-pyran-2-one, technical grade
PubChem CID 520660
SMILES O=C1CC=CCO1

Specifications

Density 1.1390g/mL
Boiling Point 103.0°C (10.0 mmHg)
Flash Point 46°C
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.139
IUPAC Name 3,6-dihydro-2H-pyran-2-one
Formula Weight 98.1
Percent Purity 90%
Chemical Name or Material 5, 6-Dihydro-2H-pyran-2-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai

RUO – Research Use Only