missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloropyridine-2-carboxylic acid, 98%

Catalog No. AAH6432003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6432003 1 g
AAH6432006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6432003 Supplier Thermo Scientific Chemicals Supplier No. H6432003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88912-24-7 | C6H3Cl2NO2 | 191.995 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88912-24-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD13185546
InChI Key QOJNAEPFSUYAFL-UHFFFAOYSA-N
Synonym 5,6-dichloropicolinic acid, 5,6-dichloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5,6-dichloro, 2,3-dichloro-6-carboxypyridine, 2-pyridinecarboxylicacid, 5,6-dichloro, acmc-209vdz, 2,3-dichloro-6-pyridinecarboxylicacid, 5,6-dichloro-pyridine-2-carboxylic acid
PubChem CID 13278016
IUPAC Name 5,6-dichloropyridine-2-carboxylic acid
SMILES C1=CC(=NC(=C1Cl)Cl)C(=O)O

Specifications

Quantity 1 g
Beilstein 4390497
Formula Weight 192
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5,6-Dichloropyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only