missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichlorobenzimidazole Hydrochloride 98.0+%, TCI America™

Catalog No. D42951G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42951G 1 g
D42955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42951G Supplier TCI America Supplier No. D42951G
Only null left

Chemical Identifiers

CAS 1087737-96-9
Molecular Formula C7H5Cl3N2
Molecular Weight (g/mol) 223.481
MDL Number MFCD16810292
InChI Key CKILENXYURTTFP-UHFFFAOYSA-N
PubChem CID 24187360
IUPAC Name 5,6-dichloro-1H-benzimidazole;hydrochloride
SMILES C1=C2C(=CC(=C1Cl)Cl)N=CN2.Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 223.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichlorobenzimidazole Hydrochloride