missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichlorobenzimidazole 1-beta-D-Ribofuranoside 98.0+%, TCI America™

Catalog No. D42921G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42921G 1 g
D4292100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42921G Supplier TCI America Supplier No. D42921G

Chemical Identifiers

CAS 53-85-0
Molecular Formula C12H12Cl2N2O4
Molecular Weight (g/mol) 319.138
MDL Number MFCD00036785
InChI Key XHSQDZXAVJRBMX-DDHJBXDOSA-N
Synonym 5,6-dichloro-1-beta-d-ribofuranosylbenzimidazole, dichlororibofuranosylbenzimidazole, drb, 5,6-dichlorobenzimidazole riboside, 5,6-dichlorobenzimidazole 1-beta-d-ribofuranoside, 5,6-dichloro-1-.beta.-d-ribofuranosylbenzimidazole, benzimidazole, 5,6-dichloro-1-beta-d-ribofuranosyl, 5,6-dichloro-1-beta-d-ribofuranosyl-1h-benzimidazole, 2r,3r,4s,5r-2-5,6-dichloro-1,3-benzodiazol-1-yl-5-hydroxymethyl oxolane-3,4-diol, 2r,3r,4s,5r-2-5,6-dichloro-1h-1,3-benzodiazol-1-yl-5-hydroxymethyl oxolane-3,4-diol
PubChem CID 5894
IUPAC Name (2R,3R,4S,5R)-2-(5,6-dichlorobenzimidazol-1-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=C2C(=CC(=C1Cl)Cl)N(C=N2)C3C(C(C(O3)CO)O)O

Specifications

Melting Point 222°C
Color White
Quantity 1 g
Formula Weight 319.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dichlorobenzimidazole 1-beta-D-Ribofuranoside
RTECSNumber DD7310000
TSCA No
Recommended Storage Freezer