missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloro-2-mercaptobenzimidazole, 98%

Catalog No. AAA1430903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1430903 1 g
AAA1430906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1430903 Supplier Thermo Scientific Chemicals Supplier No. A1430903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19462-98-7 | C7H4Cl2N2S | 219.083 g/mol

It is employed in the synthesis of 2?,3?-dideoxy and 2?,3?-unsaturated ribofuranonucleosides as potential antiviral agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 2′,3′-dideoxy and 2′,3′-unsaturated ribofuranonucleosides as potential antiviral agents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 19462-98-7
Molecular Formula C7H4Cl2N2S
Molecular Weight (g/mol) 219.083
MDL Number MFCD00052398
InChI Key AFDOMGKBKBKUHB-UHFFFAOYSA-N
PubChem CID 2774259
IUPAC Name 5,6-dichloro-1,3-dihydrobenzimidazole-2-thione
SMILES C1=C2C(=CC(=C1Cl)Cl)NC(=S)N2

Specifications

Melting Point ∼350°C (decomposition)
Odor Odorless
Quantity 1 g
Synonym 5,6-dichloro-1h-benzo d imidazole-2-thiol, 5,6-dichlorobenzimidazole-2-thiol, 5,7dicl2sh-bzind, 2-mercapto-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzimidazole-2-thiol, 5,6-dichloro-2-mercaptobenzimidazole, 5,6-dichloro-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione,5,6-dichloro-1,3-dihydro, 2h-benzimidazole-2-thione, 5,6-dichloro-1,3-dihydro, 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
Solubility Information Insoluble in water.
Formula Weight 219.09
Percent Purity 98%
Chemical Name or Material 5,6-Dichloro-2-mercaptobenzimidazole
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only