missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dibromo-1,3-benzodioxole 98.0+%, TCI America™

Catalog No. p-7128433
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47371G 1 g
D47375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47371G Supplier TCI America Supplier No. D47371G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5279-32-3
Molecular Formula C7H4Br2O2
Molecular Weight (g/mol) 279.92
MDL Number MFCD00094476
InChI Key WPYAICCSYGUFTK-UHFFFAOYSA-N
Synonym 5,6-dibromobenzo d 1,3 dioxole, 1,2-dibromo-4,5-methylenedioxy benzene, 5,6-dibromo-2h-1,3-benzodioxole, 1,2-dibromo-4,5-methylenedioxybenzene, acmc-20ap6l, 1,3-benzodioxole,5,6-dibromo, 5,6-dibromo-benzo-1,3 dioxole, 5,6-bis bromanyl-1,3-benzodioxole, 4,5-dibromo-1,2-methylenedioxybenzene, 4,5-methylenedioxy-1,2-dibromobenzene
PubChem CID 225814
IUPAC Name 5,6-dibromo-2H-1,3-benzodioxole
SMILES BrC1=CC2=C(OCO2)C=C1Br

Specifications

Melting Point 86°C
Quantity 1 g
Formula Weight 279.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Dibromo-1,3-benzodioxole