missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Diaminobenzimidazolinone 95.0+%, TCI America™
$67.11 - $195.73
Chemical Identifiers
| CAS | 55621-49-3 |
|---|---|
| Molecular Formula | C7H8N4O |
| Molecular Weight (g/mol) | 164.17 |
| MDL Number | MFCD00994340 |
| InChI Key | BRISYWKBJNSDPL-UHFFFAOYSA-N |
| Synonym | 5,6-Diamino-2-hydroxybenzimidazole |
| PubChem CID | 727371 |
| IUPAC Name | 5,6-diamino-2,3-dihydro-1H-1,3-benzodiazol-2-one |
| SMILES | NC1=C(N)C=C2NC(=O)NC2=C1 |
Chemical Identifiers
| 55621-49-3 | |
| 164.17 | |
| BRISYWKBJNSDPL-UHFFFAOYSA-N | |
| 727371 | |
| NC1=C(N)C=C2NC(=O)NC2=C1 |
| C7H8N4O | |
| MFCD00994340 | |
| 5,6-Diamino-2-hydroxybenzimidazole | |
| 5,6-diamino-2,3-dihydro-1H-1,3-benzodiazol-2-one |
Specifications
| 55621-49-3 | |
| MFCD00994340 | |
| 5,6-Diamino-2-hydroxybenzimidazole | |
| NC1=C(N)C=C2NC(=O)NC2=C1 | |
| 164.17 | |
| 164.17 | |
| Crystalline Powder |
| C7H8N4O | |
| 1 g | |
| BRISYWKBJNSDPL-UHFFFAOYSA-N | |
| 5,6-diamino-2,3-dihydro-1H-1,3-benzodiazol-2-one | |
| 727371 | |
| ≥95.0% (HPLC,T) | |
| 5,6-Diaminobenzimidazolinone |
Safety and Handling
TSCA : No