missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Diamino-1,3-diethyluracil Hydrochloride 98.0+%, TCI America™
$186.41 - $650.56
Chemical Identifiers
| CAS | 1785764-26-2 |
|---|---|
| Molecular Formula | C8H15ClN4O2 |
| Molecular Weight (g/mol) | 234.68 |
| MDL Number | MFCD22573561 |
| InChI Key | UQWLJOFXKBZWCI-UHFFFAOYSA-N |
| PubChem CID | 71299790 |
| IUPAC Name | 5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride |
| SMILES | Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O |
Chemical Identifiers
| 1785764-26-2 | |
| 234.68 | |
| UQWLJOFXKBZWCI-UHFFFAOYSA-N | |
| 5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride |
| C8H15ClN4O2 | |
| MFCD22573561 | |
| 71299790 | |
| Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O |
Specifications
| 1785764-26-2 | |
| White-Yellow | |
| MFCD22573561 | |
| UQWLJOFXKBZWCI-UHFFFAOYSA-N | |
| 5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride | |
| 71299790 | |
| ≥98.0% (HPLC,T) | |
| 5,6-Diamino-1,3-diethyluracil Hydrochloride |
| 237°C | |
| C8H15ClN4O2 | |
| 1 g | |
| Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O | |
| 234.68 | |
| 234.68 | |
| Crystalline Powder |
Safety and Handling
TSCA : No