missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diamino-1,3-diethyluracil Hydrochloride 98.0+%, TCI America™

Catalog No. D46595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46595G 5 g
D46591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46595G Supplier TCI America Supplier No. D46595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1785764-26-2
Molecular Formula C8H15ClN4O2
Molecular Weight (g/mol) 234.68
MDL Number MFCD22573561
InChI Key UQWLJOFXKBZWCI-UHFFFAOYSA-N
PubChem CID 71299790
IUPAC Name 5,6-diamino-1,3-diethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione hydrochloride
SMILES Cl.CCN1C(N)=C(N)C(=O)N(CC)C1=O

Specifications

Melting Point 237°C
Color White-Yellow
Quantity 5 g
Formula Weight 234.68
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,6-Diamino-1,3-diethyluracil Hydrochloride