missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydroquinoxaline 98.0+%, TCI America™

Catalog No. T14035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14035G 5 g
T140325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14035G Supplier TCI America Supplier No. T14035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34413-35-9
Molecular Formula C8H10N2
Molecular Weight (g/mol) 134.182
MDL Number MFCD00038078
InChI Key XCZPDOCRSYZOBI-UHFFFAOYSA-N
Synonym cyclohexapyrazine, quinoxaline, 5,6,7,8-tetrahydro, unii-m153cj9ra0, fema no. 3321, 5.6.7.8-tetrahydroquinoxaline, nutty quinoxaline, pubchem15475, acmc-209i7e, dsstox_cid_27678, dsstox_rid_82495
PubChem CID 36822
IUPAC Name 5,6,7,8-tetrahydroquinoxaline
SMILES C1CCC2=NC=CN=C2C1

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 134.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5,6,7,8-Tetrahydroquinoxaline