missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8,9,10-Hexahydrocyclohept[b]indole, 98%

Catalog No. p-7024527
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1934403 1 g
AAA1934406 5 g
2 options

Catalog No. AAA1934403

Supplier: Thermo Scientific Chemicals A1934403

Only null left

CAS: 2047-89-4 | C13H15N | 185.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2047-89-4
Molecular Formula C13H15N
Molecular Weight (g/mol) 185.27
MDL Number MFCD00101340
InChI Key XZUJMYLNFZHNLP-UHFFFAOYSA-N
Synonym 5,6,7,8,9,10-hexahydrocyclohepta b indole, 5,6,7,8,9,10-hexahydro-cyclohepta b indole, 5h,6h,7h,8h,9h,10h-cyclohepta b indole, cycloheptan a indole, 2,3-pentano-1h-indole, 5,6,7,8,9,10-hexahydrocyclohept b indole, 5,6,7,8,9,10-hexahydrocyclohepta b indole #, 6,7,8,9,10,10a-hexahydrocyclohepta b indole, 6,7,8,9,10-pentahydrocyclohepta 1,2-b indole, 6,7,8,9,10-pentahydrocyclohepta 2,1-b indole
PubChem CID 251955
IUPAC Name 5,6,7,8,9,10-hexahydrocyclohepta[b]indole
SMILES C1CCC2=C(CC1)NC3=CC=CC=C23

Specifications

Melting Point 143°C to 146°C
Quantity 1 g
Formula Weight 185.27
Percent Purity 98%
Chemical Name or Material 5,6,7,8,9,10-Hexahydrocyclohept[b]indole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only