missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene 98.0+%, TCI America™
$449.24 - $1351.44
Chemical Identifiers
| CAS | 53397-65-2 |
|---|---|
| Molecular Formula | C16H8 |
| Molecular Weight (g/mol) | 200.24 |
| InChI Key | JFYLRMKUYCIWMX-UHFFFAOYSA-N |
| Synonym | 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne |
| PubChem CID | 143044 |
| IUPAC Name | 5,6,11,12-tetradehydrodibenzo[2,1-a |
| SMILES | C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T3241100MG
|
TCI America
T3241100MG |
100 mg |
Each for $449.24
|
|
|||||
|
T3241500MG
|
TCI America
T3241500MG |
500 mg |
Each for $1,351.44
|
|
|||||
Chemical Identifiers
| 53397-65-2 | |
| 200.24 | |
| 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne | |
| 5,6,11,12-tetradehydrodibenzo[2,1-a |
| C16H8 | |
| JFYLRMKUYCIWMX-UHFFFAOYSA-N | |
| 143044 | |
| C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1 |
Specifications
| 53397-65-2 | |
| C16H8 | |
| 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctatetraene, 5,6,11,12-Tetradehydrodibenzo[a,e][8]annulene, Dibenzo[a,e]cyclooctadien-5,11-diyne | |
| C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1 | |
| 200.24 | |
| 200.24 | |
| Crystalline Powder |
| 110°C | |
| 100 mg | |
| JFYLRMKUYCIWMX-UHFFFAOYSA-N | |
| 5,6,11,12-tetradehydrodibenzo[2,1-a | |
| 143044 | |
| ≥98.0% (HPLC) | |
| 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene |
Safety and Handling
TSCA : No
Recommended Storage : Freezer