missing translation for 'onlineSavingsMsg'
Learn More

5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. p-2702527
Change view
Click to view available options
Quantity:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM09559CB 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM09559CB Supplier Thermo Scientific Chemicals Supplier No. KM09559CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 229956-98-3
Molecular Formula C8H3ClF3NO3S2
Molecular Weight (g/mol) 317.681
MDL Number MFCD00831001
InChI Key TWGONVLMWWEOFL-UHFFFAOYSA-N
Synonym 5-5-trifluoromethyl isoxazol-3-yl thiophene-2-sulfonyl chloride, 5-5-trifluoromethyl-1,2-oxazol-3-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-trifluoromethyl-3-isoxazolyl, 5-5-trifluoromethyl isoxazol-3-yl thiophene-2-sulphonyl chloride, 5-5-trifluoromethyl-isoxazol-3-yl-thiophene-2-sulfonyl chloride
PubChem CID 2777526
IUPAC Name 5-[5-(trifluoromethyl)-1,2-oxazol-3-yl]thiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1)S(=O)(=O)Cl)C2=NOC(=C2)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 317.69
Percent Purity 97%
Chemical Name or Material 5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.