missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenylhydantoin Sodium Salt 98.0+%, TCI America™
$51.98 - $398.51
Chemical Identifiers
| CAS | 630-93-3 |
|---|---|
| Molecular Formula | C15H11N2NaO2 |
| Molecular Weight (g/mol) | 274.26 |
| MDL Number | MFCD00069674,MFCD00005264 |
| InChI Key | FJPYVLNWWICYDW-UHFFFAOYSA-M |
| Synonym | Sodium 5,5-Diphenylhydantoin |
| PubChem CID | 131674077 |
| IUPAC Name | sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide |
| SMILES | [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 630-93-3 | |
| 274.26 | |
| FJPYVLNWWICYDW-UHFFFAOYSA-M | |
| 131674077 | |
| [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C15H11N2NaO2 | |
| MFCD00069674,MFCD00005264 | |
| Sodium 5,5-Diphenylhydantoin | |
| sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide |
Specifications
| White | |
| 25 g | |
| MFCD00069674,MFCD00005264 | |
| FJPYVLNWWICYDW-UHFFFAOYSA-M | |
| sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide | |
| 131674077 | |
| ≥98.0% (T) |
| Crystalline Powder | |
| C15H11N2NaO2 | |
| Sodium 5,5-Diphenylhydantoin | |
| [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 274.26 | |
| 274.25 | |
| 5,5-Diphenylhydantoin Sodium Salt |
Safety and Handling
RTECSNumber : MU1400000
TSCA : Yes