missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin Sodium Salt 98.0+%, TCI America™

Catalog No. D1331500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1331500G 500 g
D133125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1331500G Supplier TCI America Supplier No. D1331500G
Only null left

Chemical Identifiers

CAS 630-93-3
Molecular Formula C15H11N2NaO2
Molecular Weight (g/mol) 274.26
MDL Number MFCD00069674,MFCD00005264
InChI Key FJPYVLNWWICYDW-UHFFFAOYSA-M
Synonym Sodium 5,5-Diphenylhydantoin
PubChem CID 131674077
IUPAC Name sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
SMILES [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 274.25
Percent Purity ≥98.0% (T)
Chemical Name or Material 5,5-Diphenylhydantoin Sodium Salt
RTECSNumber MU1400000
TSCA Yes