missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenylhydantoin Sodium Salt 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Formula Weight | 274.25 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | 5,5-Diphenylhydantoin Sodium Salt |
Safety and Handling
| RTECSNumber | MU1400000 |
| TSCA | Yes |