missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenylhydantoin sodium salt, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 295°C to 298°C |
| Color | White |
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 99% |
| Merck Index | 15,7433 |
| Solubility Information | Solubility in water: practically insoluble |
| IUPAC Name | sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. May cause an allergic skin reaction. Suspected of causing cancer. Suspected of damaging fertility. Suspected of damaging the unborn child. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: G |
| EINECSNumber | 211-148-2 |
RUO – Research Use Only