Learn More
5,5-Diphenylhydantoin, 99%
CAS: 57-41-0 | C15H12N2O2 | 252.273 g/mol
Supplier: Thermo Scientific Chemicals A1701336
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 5,5-Diphenylhydantoin | |
| 293°C to 298°C | |
| MFCD00005264 | |
| 384532 | |
| phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan | |
| C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3 | |
| 252.273 | |
| CHEBI:8107 | |
| 99% |
| 57-41-0 | |
| C15H12N2O2 | |
| 500 g | |
| 14,7322 | |
| CXOFVDLJLONNDW-UHFFFAOYSA-N | |
| 5,5-diphenylimidazolidine-2,4-dione | |
| 1775 | |
| 252.27 |
Chemical Identifiers
| 57-41-0 | |
| 252.273 | |
| CXOFVDLJLONNDW-UHFFFAOYSA-N | |
| 1775 | |
| 5,5-diphenylimidazolidine-2,4-dione |
| C15H12N2O2 | |
| MFCD00005264 | |
| phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan | |
| CHEBI:8107 | |
| C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3 |
Safety and Handling
GHS H Statement
H350-H360-H317
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350-H361d-H500
EINECSNumber : 200-328-6
RTECSNumber : MU1050000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only