missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenylhydantoin 99.0+%, TCI America™
$94.33 - $94.33
Chemical Identifiers
| CAS | 57-41-0 |
|---|---|
| Molecular Formula | C15H12N2O2 |
| Molecular Weight (g/mol) | 252.273 |
| MDL Number | MFCD00005264 |
| InChI Key | CXOFVDLJLONNDW-UHFFFAOYSA-N |
| Synonym | phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan |
| PubChem CID | 1775 |
| ChEBI | CHEBI:8107 |
| IUPAC Name | 5,5-diphenylimidazolidine-2,4-dione |
| SMILES | C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3 |
Chemical Identifiers
| 57-41-0 | |
| 252.273 | |
| CXOFVDLJLONNDW-UHFFFAOYSA-N | |
| 1775 | |
| 5,5-diphenylimidazolidine-2,4-dione |
| C15H12N2O2 | |
| MFCD00005264 | |
| phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan | |
| CHEBI:8107 | |
| C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3 |
Specifications
| 57-41-0 | |
| White | |
| MFCD00005264 | |
| phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan | |
| C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3 | |
| 252.273 | |
| CHEBI:8107 | |
| ≥99.0% (T) | |
| 5,5-Diphenylhydantoin |
| 298°C | |
| C15H12N2O2 | |
| 25 g | |
| CXOFVDLJLONNDW-UHFFFAOYSA-N | |
| 5,5-diphenylimidazolidine-2,4-dione | |
| 1775 | |
| 252.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (9)-0621
RTECSNumber : MU1050000
TSCA : No