missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin 99.0+%, TCI America™

Catalog No. D089425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D089425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D089425G Supplier TCI America Supplier No. D089425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

Melting Point 298°C
Color White
Quantity 25 g
Formula Weight 252.27
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Diphenylhydantoin
EINECSNumber (9)-0621
RTECSNumber MU1050000
TSCA No