missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethylhydantoin 98.0+%, TCI America™

Catalog No. p-7126408
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D073925G 25 g
D0739500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D073925G Supplier TCI America Supplier No. D073925G
Only null left

Chemical Identifiers

CAS 77-71-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005266
InChI Key YIROYDNZEPTFOL-UHFFFAOYSA-N
Synonym 5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin
PubChem CID 6491
IUPAC Name 5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)NC(=O)NC1=O

Specifications

Melting Point 175°C
Color White
Quantity 25 g
Formula Weight 128.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5-Dimethylhydantoin
EINECSNumber (5)-0441
RTECSNumber MU0977000
TSCA Yes