missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethylhydantoin, 97%

Catalog No. p-7043903
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0338822 100 g
AAL0338836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0338822 Supplier Thermo Scientific Chemicals Supplier No. L0338822
Only null left

CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-71-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005266
InChI Key YIROYDNZEPTFOL-UHFFFAOYSA-N
Synonym 5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin
PubChem CID 6491
IUPAC Name 5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)NC(=O)NC1=O

Specifications

Melting Point 174°C to 178°C
Flash Point 193°C (379°F)
Odor Odorless
Quantity 100 g
Beilstein 2827
Formula Weight 128.13
Percent Purity 97%
Chemical Name or Material 5,5-Dimethylhydantoin
EINECSNumber 201-051-3
RTECSNumber MU0977000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only