missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane 98.0+%, TCI America™
$201.32 - $603.95
Chemical Identifiers
| CAS | 5123-13-7 |
|---|---|
| Molecular Formula | C11H15BO2 |
| Molecular Weight (g/mol) | 190.05 |
| MDL Number | MFCD05663851 |
| InChI Key | FQMSNYZDFMWLGC-UHFFFAOYSA-N |
| Synonym | (5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzene, Phenylboronic Acid Neopentyl Glycol Ester |
| PubChem CID | 11788741 |
| IUPAC Name | 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane |
| SMILES | CC1(C)COB(OC1)C1=CC=CC=C1 |
Chemical Identifiers
| 5123-13-7 | |
| 190.05 | |
| FQMSNYZDFMWLGC-UHFFFAOYSA-N | |
| 11788741 | |
| CC1(C)COB(OC1)C1=CC=CC=C1 |
| C11H15BO2 | |
| MFCD05663851 | |
| (5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzene, Phenylboronic Acid Neopentyl Glycol Ester | |
| 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane |
Specifications
| 5123-13-7 | |
| White | |
| MFCD05663851 | |
| (5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzene, Phenylboronic Acid Neopentyl Glycol Ester | |
| CC1(C)COB(OC1)C1=CC=CC=C1 | |
| 190.05 | |
| 190.05 | |
| Crystalline Powder |
| 65°C | |
| C11H15BO2 | |
| 5 g | |
| FQMSNYZDFMWLGC-UHFFFAOYSA-N | |
| 5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane | |
| 11788741 | |
| ≥98.0% (GC,T) | |
| 5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator