missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5'-Dimethyl-2,2'-bipyridyl 98.0+%, TCI America™
$76.43 - $654.29
Chemical Identifiers
| CAS | 1762-34-1 |
|---|---|
| Molecular Formula | C12H12N2 |
| Molecular Weight (g/mol) | 184.242 |
| MDL Number | MFCD01740554 |
| InChI Key | PTRATZCAGVBFIQ-UHFFFAOYSA-N |
| Synonym | 5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl |
| PubChem CID | 15664 |
| IUPAC Name | 5-methyl-2-(5-methylpyridin-2-yl)pyridine |
| SMILES | CC1=CN=C(C=C1)C2=NC=C(C=C2)C |
Chemical Identifiers
| 1762-34-1 | |
| 184.242 | |
| PTRATZCAGVBFIQ-UHFFFAOYSA-N | |
| 15664 | |
| CC1=CN=C(C=C1)C2=NC=C(C=C2)C |
| C12H12N2 | |
| MFCD01740554 | |
| 5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl | |
| 5-methyl-2-(5-methylpyridin-2-yl)pyridine |
Specifications
| 1762-34-1 | |
| C12H12N2 | |
| 1 g | |
| PTRATZCAGVBFIQ-UHFFFAOYSA-N | |
| 5-methyl-2-(5-methylpyridin-2-yl)pyridine | |
| 15664 | |
| ≥98.0% (GC,T) | |
| 5,5′-Dimethyl-2,2′-bipyridyl |
| 115°C | |
| MFCD01740554 | |
| 5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl | |
| CC1=CN=C(C=C1)C2=NC=C(C=C2)C | |
| 184.242 | |
| 184.24 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DW1766000
TSCA : No