missing translation for 'onlineSavingsMsg'
Learn More

5,5‴-Dihexyl-2,2':5',2″:5″,2‴-quaterthio 98+%, TCI America™

Catalog No. D4842100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4842100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4842100MG Supplier TCI America Supplier No. D4842100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132814-92-7
Molecular Formula C28H34S4
Molecular Weight (g/mol) 498.82
MDL Number MFCD21396218
InChI Key KGJQHEPGNCWZRN-UHFFFAOYSA-N
Synonym alpha,omega-Dihexylquaterthiophene
PubChem CID 10720155
IUPAC Name 2-hexyl-5-[5-[5-(5-hexylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene
SMILES CCCCCCC1=CC=C(S1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)CCCCCC

Specifications

Melting Point 180°C
Quantity 100 mg
Formula Weight 498.82
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′′′-Dihexyl-2,2′:5′,2′′:5′′,2′′