missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-4,4'-dinonyl-2,2'-bithiazole 97.0+%, TCI America™

Catalog No. d4558200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4558200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4558200MG Supplier TCI America Supplier No. D4558200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 172100-44-6
Molecular Formula C24H38Br2N2S2
Molecular Weight (g/mol) 578.51
InChI Key NVYFBJQBTRFBCJ-UHFFFAOYSA-N
PubChem CID 19437292
IUPAC Name 5-bromo-2-(5-bromo-4-nonyl-1,3-thiazol-2-yl)-4-nonyl-1,3-thiazole
SMILES CCCCCCCCCC1=C(SC(=N1)C2=NC(=C(S2)Br)CCCCCCCCC)Br

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 578.51
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Dibromo-4,4′-dinonyl-2,2′-bithiazole