missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene 95.0+%, TCI America™

Catalog No. D41841G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41841G 1 g
D4184200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41841G Supplier TCI America Supplier No. D41841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 214493-03-5
Molecular Formula C20H28Br2S2
Molecular Weight (g/mol) 492.37
MDL Number MFCD28386098
InChI Key MWUMRWQISLASDX-UHFFFAOYSA-N
PubChem CID 11953640
IUPAC Name 5,5'-dibromo-4,4'-dihexyl-2,2'-bithiophene
SMILES CCCCCCC1=C(Br)SC(=C1)C1=CC(CCCCCC)=C(Br)S1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 492.37
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5,5′-Dibromo-4,4′-dihexyl-2,2′-bithiophene
TSCA No
Recommended Storage Refrigerator