missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-2,2'-bithiophene 98.0+%, TCI America™

Catalog No. D27551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27551G 1 g
D27555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27551G Supplier TCI America Supplier No. D27551G

Chemical Identifiers

CAS 4805-22-5
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.048
MDL Number MFCD00219110
InChI Key SXNCMLQAQIGJDO-UHFFFAOYSA-N
Synonym 5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4r
PubChem CID 638908
IUPAC Name 2-bromo-5-(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)Br)C2=CC=C(S2)Br

Specifications

Melting Point 146°C
Quantity 1 g
Formula Weight 324.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Dibromo-2,2′-bithiophene