missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-2,2'-bithiophene, 98%

Catalog No. AAH5633706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5633706 5 g
AAH5633714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5633706 Supplier Thermo Scientific Chemicals Supplier No. H5633706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4805-22-5 | C8H4Br2S2 | 324.048 g/mol

It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C.

Chemical Identifiers

CAS 4805-22-5
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.048
MDL Number MFCD00219110
InChI Key SXNCMLQAQIGJDO-UHFFFAOYSA-N
Synonym 5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4r
PubChem CID 638908
IUPAC Name 2-bromo-5-(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)Br)C2=CC=C(S2)Br

Specifications

Melting Point 144°C to 146°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 324.06
Percent Purity 99%
Chemical Name or Material 5,5'-Dibromo-2,2'-bithiophene
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only