missing translation for 'onlineSavingsMsg'
Learn More

5,5″-Dibromo-2,2':5',2″-terthiophene 97.0+%, TCI America™

Catalog No. D40501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40501G 1 g
D40505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40501G Supplier TCI America Supplier No. D40501G

Chemical Identifiers

CAS 98057-08-0
Molecular Formula C12H6Br2S3
Molecular Weight (g/mol) 406.168
MDL Number MFCD00219109
InChI Key KXFPYYJGYVYXIB-UHFFFAOYSA-N
Synonym 2,5-Bis(5-bromo-2-thienyl)thiophene
PubChem CID 176690
IUPAC Name 2,5-bis(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)C2=CC=C(S2)Br)C3=CC=C(S3)Br

Specifications

Melting Point 158°C
Color Yellow
Quantity 1 g
Formula Weight 406.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′′-Dibromo-2,2′:5′,2′′-terthiophene