missing translation for 'onlineSavingsMsg'
Learn More

5,5″-Bis(trimethylstannyl)-2,2':5',2″-terthiophene 98.0+%, TCI America™

Catalog No. B5690200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5690200MG 200 mg
B56901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5690200MG Supplier TCI America Supplier No. B5690200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 178931-63-0
Molecular Formula C18H24S3Sn2
Molecular Weight (g/mol) 573.99
MDL Number MFCD30489177
InChI Key FIKQPMKITUNTEK-UHFFFAOYSA-N
PubChem CID 71353375
IUPAC Name trimethyl-[5-[5-(5-trimethylstannylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]stannane
SMILES C[Sn](C)(C)C1=CC=C(S1)C2=CC=C(S2)C3=CC=C(S3)[Sn](C)(C)C

Specifications

Melting Point 122°C
Quantity 200 mg
UN Number 3146
Formula Weight 573.99
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′′-Bis(trimethylstannyl)-2,2′:5′,2′′-terthiophene
TSCA No
Recommended Storage Refrigerator