missing translation for 'onlineSavingsMsg'
Learn More

5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid, 97%, Thermo Scientific™

Catalog No. CC27101CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27101CB 250mg Amber Glass Bottle
CC27101DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC27101CB Supplier Thermo Scientific Chemicals Supplier No. CC27101CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103031-30-7
Molecular Formula C15H20O2
Molecular Weight (g/mol) 232.323
MDL Number MFCD04115382
InChI Key KSEZYZSBKCPEKP-UHFFFAOYSA-N
Synonym 5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl, 2-naphthalenecarboxylicacid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl, acmc-1c8hs, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthoic acid, 5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthoic acid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylicacid, 5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-naphthalene-2-carboxylic acid
PubChem CID 227638
IUPAC Name 5,5,8,8-tetramethyl-6,7-dihydronaphthalene-2-carboxylic acid
SMILES CC1(CCC(C2=C1C=CC(=C2)C(=O)O)(C)C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 232.32
Percent Purity 97%
Chemical Name or Material 5,5,8,8-Tetramethyl-5,6,7,8-tetrahydro-2-naphthalenecarboxylic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.