missing translation for 'onlineSavingsMsg'
Learn More

5,5',7-Indigotrisulfonic Acid Potassium Salt, Spectrum™ Chemical

Catalog No. 18608279
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608279 5 g Amber Glass Bottle
18608278 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608279 Supplier Spectrum Chemical Mfg Cor Supplier No. I11915GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67627-18-3

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Molecular Weight (g/mol) 616.71
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 5,5',7-Indigotrisulfonic Acid Potassium Salt

CAUTION: For use by qualified personnel for research and development only. Not for food or drug use. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.