missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Pyridyl)-1H-tetrazole, 98%, Thermo Scientific Chemicals

$126.93 - $126.93

Chemical Identifiers

CAS 14389-12-9
Molecular Formula C6H5N5
Molecular Weight (g/mol) 147.141
MDL Number MFCD00068115
InChI Key JMILUUVWLRKJFB-UHFFFAOYSA-N
Synonym 5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
PubChem CID 482249
IUPAC Name 4-(2H-tetrazol-5-yl)pyridine
SMILES C1=CN=CC=C1C2=NNN=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2074103
View Documents
Thermo Scientific Chemicals
B2074103
1 g
Each for $126.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14389-12-9
147.141
JMILUUVWLRKJFB-UHFFFAOYSA-N
482249
C1=CN=CC=C1C2=NNN=N2
C6H5N5
MFCD00068115
5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
4-(2H-tetrazol-5-yl)pyridine

Specifications

14389-12-9
MFCD00068115
5-4-pyridyl-1h-tetrazole, 4-2h-tetrazol-5-yl pyridine, 4-1h-tetrazol-5-yl pyridine, 4-2h-tetrazol-5-yl-pyridine, 4-1h-1,2,3,4-tetrazol-5-yl pyridine, 4-2h-1,2,3,4-tetrazol-5-yl pyridine, pyridine, 4-2h-tetrazol-5-yl, 5-4-pyridyl-1 h-tetrazole, 5-4-pyridyl-2h-1,2,3,4-tetraazole, acmc-20amxi
C1=CN=CC=C1C2=NNN=N2
147.141
147.14
5-(4-Pyridyl)-1H-tetrazole
C6H5N5
1 g
JMILUUVWLRKJFB-UHFFFAOYSA-N
4-(2H-tetrazol-5-yl)pyridine
482249
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only