missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. h15501g
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
H15501G 1 g
H1550200MG 200 mg
2 options

Catalog No. H15501G

Supplier: TCI America H15501G

Only null left

Chemical Identifiers

CAS 207290-72-0
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.279
MDL Number MFCD03425728
InChI Key QGNLQHONMNFZJP-UHFFFAOYSA-N
Synonym 5-(4-Hydroxypiperidino)thiophene-2-carboxaldehyde
PubChem CID 2763540
IUPAC Name 5-(4-hydroxypiperidin-1-yl)thiophene-2-carbaldehyde
SMILES C1CN(CCC1O)C2=CC=C(S2)C=O

Specifications

Melting Point 145°C
Quantity 1 g
Formula Weight 211.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde