missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 145°C |
| Quantity | 200 mg |
| Formula Weight | 211.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde |
Safety and Handling
| TSCA | No |