missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)valeric acid, 97%

Catalog No. p-7047188
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL17935MD 250 mg
AAL1793503 1 g
AAL1793506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL17935MD Supplier Thermo Scientific Chemicals Supplier No. L17935MD
Only null left

CAS: 24484-22-8 | C11H12FO2 | 195.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24484-22-8
Molecular Formula C11H12FO2
Molecular Weight (g/mol) 195.21
MDL Number MFCD00800242
InChI Key BXEFPLJKYWEEAN-UHFFFAOYSA-M
PubChem CID 2734670
IUPAC Name 5-(4-fluorophenyl)pentanoic acid
SMILES [O-]C(=O)CCCCC1=CC=C(F)C=C1

Specifications

Melting Point 73°C to 75°C
Quantity 250 mg
Beilstein 7918899
Synonym 5-4-fluorophenyl valeric acid, 5-4-fluorophenyl pentanoic acid, benzenepentanoic acid, 4-fluoro, 5-4'-fluorophenyl valeric acid, acmc-1cj6r, benzenepentanoic acid,4-fluoro, 5-4-fluorophenyl-pentanoic acid, 5-4-fluoro-phenyl-pentanoic acid
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 5-(4-Fluorophenyl)valeric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only